Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055628.1 | XP_017911117.1 | 2 - 3 | 893 | 62 | |
GenBank mRNA | JO646145.2 | - | 2 - 3 | 958 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Capra hircus Probe spans exons |