Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967923.2 | XP_013823377.2 | 4 - 5 | 493 | 97 | |
GenBank mRNA | JO583040.1 | - | 1 - 2 | 512 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |