Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062512.1 | XP_017918001.1 | 3 - 4 | 483 | 69 | |
GenBank mRNA | KA327169.1 | - | 3 - 4 | 518 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |