Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054666.1 | XP_017910155.1 | 1 - 2 | 1776 | 80 | |
GenBank mRNA | JO616559.2 | - | 1 - 2 | 181 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |