Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054666.1 | XP_017910155.1 | 6 - 7 | 2982 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |