Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFFO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054762.1 | XP_017910251.1 | 5 - 6 | 1112 | 61 | |
XM_018054763.1 | XP_017910252.1 | 3 - 4 | 893 | 61 | ||
GenBank mRNA | JO346228.2 | - | 2 - 3 | 209 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |