Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678129.3 | XP_005678186.3 | 1 - 2 | 248 | 67 | |
GenBank mRNA | JO508413.2 | - | 3 - 4 | 608 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |