Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682964.2 | XP_005683021.2 | 9 - 10 | 2478 | 61 | |
XM_005682965.2 | XP_005683022.2 | 8 - 9 | 2284 | 61 | ||
XM_005682966.2 | XP_005683023.2 | 7 - 8 | 2119 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |