Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696031.3 | XP_005696088.3 | 1 - 2 | 610 | 68 | |
GenBank mRNA | JO403980.2 | - | 1 - 2 | 1442 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |