Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R14D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054436.1 | XP_017909925.1 | 5 - 6 | 582 | 102 | |
XM_018054437.1 | XP_017909926.1 | 4 - 5 | 439 | 102 | ||
GenBank mRNA | JT711150.1 | - | 3 - 4 | 792 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |