Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VKORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040031.1 | XP_017895520.1 | 2 - 3 | 342 | 91 | |
GenBank mRNA | JO577977.1 | - | 1 - 2 | 891 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |