Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050331.1 | XP_017905820.1 | 4 - 5 | 1342 | 75 | |
GenBank mRNA | JT639803.1 | - | 4 - 5 | 1145 | 75 | |
JT759829.1 | - | 3 - 4 | 1069 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |