Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684859.3 | XP_005684916.2 | 7 - 8 | 6338 | 83 | |
XM_018053281.1 | XP_017908770.1 | 9 - 10 | 6580 | 83 | ||
XM_018053282.1 | XP_017908771.1 | 9 - 10 | 6398 | 83 | ||
XM_018053283.1 | XP_017908772.1 | 9 - 10 | 6580 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |