Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695351.3 | XP_005695408.2 | 2 - 3 | 887 | 81 | |
XM_005695352.3 | XP_005695409.2 | 2 - 3 | 878 | 81 | ||
GenBank mRNA | JO421733.2 | - | 3 - 4 | 776 | 81 | |
JO434683.2 | - | 3 - 4 | 730 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |