Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696760.3 | XP_005696817.1 | 3 - 4 | 652 | 95 | |
XM_005696762.3 | XP_005696819.1 | 4 - 5 | 698 | 95 | ||
XM_018039208.1 | XP_017894697.1 | 3 - 4 | 762 | 95 | ||
XM_018039209.1 | XP_017894698.1 | 3 - 4 | 757 | 95 | ||
GenBank mRNA | JI861635.1 | - | 2 - 3 | 493 | 95 | |
JO386614.2 | - | 1 - 2 | 225 | 95 | ||
JO576367.1 | - | 3 - 4 | 794 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |