Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BGRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700586.3 | XP_005700643.1 | 2 - 3 | 473 | 82 | |
XM_018043954.1 | XP_017899443.1 | 2 - 3 | 473 | 82 | ||
GenBank mRNA | JO498150.2 | - | 2 - 3 | 543 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |