Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972619.2 | XP_013828073.2 | 11 - 12 | 2186 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |