Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038287.1 | XP_017893776.1 | 2 - 3 | 350 | 66 | |
GenBank mRNA | JO440171.2 | - | 2 - 3 | 316 | 66 | |
JT673942.1 | - | 1 - 2 | 243 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |