Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013974522.2 | XP_013829976.1 | 2 - 3 | 324 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |