Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677143.3 | XP_005677200.2 | 5 - 6 | 849 | 79 | |
GenBank mRNA | JO612023.2 | - | 2 - 3 | 1290 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |