Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680858.1 | XP_005680915.1 | 4 - 5 | 639 | 106 | |
GenBank mRNA | JO417360.2 | - | 2 - 3 | 185 | 106 | |
JO612746.2 | - | 4 - 5 | 643 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |