Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H2orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686840.2 | XP_005686897.1 | 3 - 4 | 1167 | 93 | |
XM_005686841.3 | XP_005686898.1 | 4 - 5 | 3829 | 93 | ||
XM_013967789.2 | XP_013823243.1 | 3 - 4 | 1188 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |