Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H2orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686840.2 | XP_005686897.1 | 7 - 8 | 1599 | 67 | |
XM_005686841.3 | XP_005686898.1 | 8 - 9 | 4261 | 67 | ||
XM_013967789.2 | XP_013823243.1 | 7 - 8 | 1620 | 67 | ||
GenBank mRNA | JT701341.1 | - | 7 - 8 | 1177 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |