Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063681.1 | XP_017919170.1 | 7 - 8 | 1418 | 54 | |
XM_018063682.1 | XP_017919171.1 | 7 - 8 | 1433 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |