Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676158.2 | XP_005676215.2 | 3 - 4 | 411 | 67 | |
GenBank mRNA | JO578197.1 | - | 1 - 2 | 950 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |