Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049580.1 | XP_017905069.1 | 4 - 5 | 631 | 100 | |
XM_018049581.1 | XP_017905070.1 | 4 - 5 | 611 | 100 | ||
GenBank mRNA | JO518901.2 | - | 1 - 2 | 115 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |