Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H19orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062416.1 | XP_017917905.1 | 7 - 8 | 742 | 73 | |
XM_018062417.1 | XP_017917906.1 | 7 - 8 | 726 | 73 | ||
XM_018062418.1 | XP_017917907.1 | 7 - 8 | 742 | 73 | ||
XM_018062419.1 | XP_017917908.1 | 7 - 8 | 739 | 73 | ||
GenBank mRNA | JO617105.2 | - | 3 - 4 | 481 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |