Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050581.1 | XP_017906070.1 | 3 - 4 | 1151 | 87 | |
XM_018050582.1 | XP_017906071.1 | 3 - 4 | 1094 | 87 | ||
XM_018050583.1 | XP_017906072.1 | 3 - 4 | 1149 | 87 | ||
GenBank mRNA | KA351871.1 | - | 3 - 4 | 1118 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |