Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD21L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688329.2 | XP_005688386.1 | 7 - 8 | 1212 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |