Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693795.3 | XP_005693852.2 | 5 - 6 | 1041 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |