Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064095.1 | XP_017919584.1 | 2 - 3 | 1689 | 53 | |
GenBank mRNA | JO425897.1 | - | 9 - 10 | 1863 | 53 | |
JO595834.2 | - | 2 - 3 | 1627 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |