Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039478.1 | XP_017894967.1 | 2 - 3 | 319 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |