Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046213.1 | XP_017901702.1 | 9 - 10 | 1507 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |