Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680753.3 | XP_005680810.1 | 1 - 2 | 422 | 106 | |
GenBank mRNA | JO356473.2 | - | 2 - 3 | 328 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |