Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013969002.2 | XP_013824456.1 | 2 - 3 | 1511 | 97 | |
XM_018064344.1 | XP_017919833.1 | 2 - 3 | 1630 | 97 | ||
XM_018064349.1 | XP_017919838.1 | 2 - 3 | 1254 | 97 | ||
GenBank mRNA | JO375717.2 | - | 2 - 3 | 672 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |