Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058059.1 | XP_017913548.1 | 2 - 3 | 382 | 77 | |
XM_018058061.1 | XP_017913550.1 | 2 - 3 | 382 | 77 | ||
GenBank mRNA | KA348401.1 | - | 4 - 5 | 1168 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |