Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JHY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689564.3 | XP_005689621.2 | 4 - 5 | 1152 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |