Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042570.1 | XP_017898059.1 | 10 - 11 | 2702 | 59 | |
XM_018042572.1 | XP_017898061.1 | 9 - 10 | 2678 | 59 | ||
XM_018042573.1 | XP_017898062.1 | 10 - 11 | 2552 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |