Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043708.1 | XP_017899197.1 | 3 - 4 | 377 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |