Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051121.1 | XP_017906610.1 | 7 - 8 | 915 | 58 | |
GenBank mRNA | JT634712.1 | - | 2 - 3 | 640 | 58 | |
JT713842.1 | - | 1 - 2 | 640 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |