Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHNAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042683.1 | XP_017898172.1 | 3 - 4 | 439 | 106 | |
XM_018042684.1 | XP_017898173.1 | 3 - 4 | 439 | 106 | ||
XM_018042686.1 | XP_017898175.1 | 3 - 4 | 439 | 106 | ||
XM_018042687.1 | XP_017898176.1 | 3 - 4 | 441 | 106 | ||
XM_018042688.1 | XP_017898177.1 | 3 - 4 | 440 | 106 | ||
XM_018042689.1 | XP_017898178.1 | 3 - 4 | 450 | 106 | ||
GenBank mRNA | KA347340.1 | - | 3 - 4 | 423 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |