Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676331.3 | XP_005676388.1 | 4 - 5 | 939 | 79 | |
GenBank mRNA | JT677816.1 | - | 4 - 5 | 804 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |