Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H19orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965535.2 | XP_013820989.1 | 2 - 3 | 496 | 72 | |
GenBank mRNA | JO597976.2 | - | 2 - 3 | 407 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |