Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054631.1 | XP_017910120.1 | 4 - 5 | 764 | 68 | |
GenBank mRNA | KA336498.1 | - | 2 - 3 | 541 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |