Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHURC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685980.3 | XP_005686037.1 | 1 - 2 | 139 | 83 | |
XM_005685981.3 | XP_005686038.1 | 1 - 2 | 139 | 83 | ||
XM_013967241.2 | XP_013822695.1 | 1 - 2 | 139 | 83 | ||
GenBank mRNA | JO453599.2 | - | 3 - 4 | 491 | 83 | |
JO613218.2 | - | 3 - 4 | 2704 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |