Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697710.3 | XP_005697767.1 | 6 - 7 | 1058 | 73 | |
GenBank mRNA | JT691206.1 | - | 13 - 14 | 4338 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |