Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013968700.2 | XP_013824154.1 | 1 - 2 | 1112 | 70 | |
GenBank mRNA | JO430138.2 | - | 1 - 2 | 1413 | 70 | |
JT677400.1 | - | 1 - 2 | 654 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |