Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058505.1 | XP_017913994.1 | 2 - 3 | 1089 | 90 | |
GenBank mRNA | JO638347.2 | - | 2 - 3 | 336 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |