Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058399.1 | XP_017913888.1 | 2 - 3 | 443 | 93 | |
GenBank mRNA | JO511566.2 | - | 2 - 3 | 129 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |