Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696261.3 | XP_005696318.3 | 12 - 13 | 2223 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |